Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ S 07662
Inverse agonist of the constitutive androstane receptor (CAR)
Specifications
Purity | >98% |
---|---|
Product Type | S 07662 |
Molecular Weight (g/mol) | 300.38 |
Inhibitors | S 07662 |
Description
Inverse agonist of the constitutive androstane receptor (CAR). Inhibits CAR activity via recruitment of nuclear receptor co-repressor (NCoR). Suppresses CITCO -induced CYP2B6 gene expression in human primary hepatocytes.Specifications
>98% | |
S 07662 | |
300.38 | |
S 07662 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title