Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-1-BOC-3-(Hydroxymethyl)piperidine, 97%, Thermo Scientific™

Catalog No. AC437480050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 140695-84-7
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.29
InChI Key OJCLHERKFHHUTB-VIFPVBQESA-N
Synonym s-1-boc-3-hydroxymethyl piperidine, s-tert-butyl 3-hydroxymethyl piperidine-1-carboxylate, s-n-boc-3-piperidinemethanol, tert-butyl 3s-3-hydroxymethyl piperidine-1-carboxylate, s-1-n-boc-3-hydroxymethyl-piperidine, s-1-boc-3-hyroxymethyl piperidine, s-1-n-boc-3-hydroxymethyl piperidine, s-n-boc-3-hyroxymethyl piperidine, s-1-boc-3-hydroxymethylpiperidine
PubChem CID 1514448
IUPAC Name tert-butyl (3S)-3-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(C1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 92.3°C to 94.2°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation 15
Formula Weight 215.29
Percent Purity 97%
Physical Form Powder
Chemical Name or Material (S)-1-BOC-3-(Hydroxymethyl)piperidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.