Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-2-(Aminomethyl)-1-ethylpyrrolidine, 99%

Catalog No. p-2689305
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 22795-99-9 | C7H16N2 | 128.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22795-99-9
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD00191371
InChI Key UNRBEYYLYRXYCG-ZETCQYMHSA-N
Synonym s-1-ethylpyrrolidin-2-yl methanamine, s---2-aminomethyl-1-ethylpyrrolidine, s-2-aminomethyl-1-ethylpyrrolidine, 2s-1-ethylpyrrolidin-2-yl methanamine, s-1-ethyl-2-aminomethylpyrrolidine, 2s-2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl-, 2s, pubchem18590
PubChem CID 643457
IUPAC Name [(2S)-1-ethylpyrrolidin-2-yl]methanamine
SMILES CCN1CCCC1CN

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9190g/mL
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4660 to 1.4680
Quantity 1 g
Specific Gravity 0.919
Specific Rotation Condition − 107.00 (20.00°C c=1,methanol)
Specific Rotation − 107.00
Formula Weight 128.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material (S)-2-(Aminomethyl)-1-ethylpyrrolidine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.