Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methoxypropanol, 97%, Acros Organics

Click to view available options
Quantity:
1mL
2.5mL
10mL
Packaging:
Glass bottle
116422-39-0
C4H10O2
90.12
MFCD01632593
YTTFFPATQICAQN-BYPYZUCNSA-N
2s-2-methoxypropan-1-ol,s-+-2-methoxypropanol,s-2-methoxypropan-1-ol,unii-6725gr28fg,2-methoxy-1-propanol, 2s,1-propanol, 2-methoxy-, 2s,2s-2-methoxy-1-propanol,s-2-methoxy-1-propanol,2-s-methoxypropan-1-ol,1-propanol, 2-methoxy-, s
642402
(2S)-2-methoxypropan-1-ol
CC(CO)OC
116422-39-0
C4H10O2
90.12
MFCD01632593
YTTFFPATQICAQN-BYPYZUCNSA-N
2s-2-methoxypropan-1-ol,s-+-2-methoxypropanol,s-2-methoxypropan-1-ol,unii-6725gr28fg,2-methoxy-1-propanol, 2s,1-propanol, 2-methoxy-, 2s,2s-2-methoxy-1-propanol,s-2-methoxy-1-propanol,2-s-methoxypropan-1-ol,1-propanol, 2-methoxy-, s
642402
(2S)-2-methoxypropan-1-ol
CC(CO)OC
Infrared Spectrum Authentic
Density 0.9380g/mL
Specific Rotation Condition + 39.20 (20.00°C c=1.66,CHCl3)
Specific Gravity 0.938
Flash Point 33°C
Water 0.1% max. (K.F.)
Refractive Index 1.4060 to 1.4080
Quantity 1mL
Specific Rotation + 39.20
Color Colorless
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula CH3CH(OCH3)CH2OH
Formula Weight 90.12
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Methoxypropanol, 99%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

;GHS P StatementKeep away from heat/sparks/open flames/hot surfaces.- No smoking

TSCA : TSCA