Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-2-Methoxypropanol, 97%

Catalog No. p-158274
Click to view available options
Quantity:
1 mL
2.5 mL
10 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 116422-39-0 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116422-39-0
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD01632593
InChI Key YTTFFPATQICAQN-BYPYZUCNSA-N
Synonym 2s-2-methoxypropan-1-ol, s-+-2-methoxypropanol, s-2-methoxypropan-1-ol, unii-6725gr28fg, 2-methoxy-1-propanol, 2s, 1-propanol, 2-methoxy-, 2s, 2s-2-methoxy-1-propanol, s-2-methoxy-1-propanol, 2-s-methoxypropan-1-ol, 1-propanol, 2-methoxy-, s
PubChem CID 642402
IUPAC Name (2S)-2-methoxypropan-1-ol
SMILES CC(CO)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9380g/mL
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4060 to 1.4080
Linear Formula CH3CH(OCH3)CH2OH
Quantity 1 mL
Specific Gravity 0.938
Specific Rotation Condition + 39.20 (20.00°C c=1.66,CHCl3)
Specific Rotation + 39.20
Formula Weight 90.12
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (S)-(+)-2-Methoxypropanol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.