Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(S)-3-Aminotetrahydrofuran hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
| Melting Point | 140.0°C to 145.0°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 94.0 to max. 106% min. (Argentometry) |
| Quantity | 5 g |
| Specific Rotation | -7.4 |
| Formula Weight | 123.58 |
| Percent Purity | 95% |
| Chemical Name or Material | (S)-3-Aminotetrahydrofuran hydrochloride |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Call a POISON CENTER or doctor/physician if you feel unwell. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction