Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea Methanesulfonate 98.0+%, TCI America™
SDP

Catalog No. N096610MG Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 mg
50 mg

Chemical Identifiers

CAS 182004-65-5
Molecular Formula C17H21N3O6S2
Molecular Weight (g/mol) 427.49
MDL Number MFCD00952138
InChI Key WGIKEBHIKKWJLG-UHFFFAOYSA-N
Synonym kb-r7943 mesylate, 2-2-4-4-nitrobenzyloxy phenyl ethyl isothiourea mesylate, 4-4-nitrobenzyl oxy phenethyl carbamimidothioate methanesulfonate, s-4-4-nitrobenzyl oxy phenethyl isothiourea methanesulfonate, 2-2-4-4-nitrobenzyloxy phenyl ethyl isothiourea, methane sulfonate, 2-4-4-nitrophenyl methoxy phenyl ethyl sulfanylmethanimidamide; methanesulfonic acid, d0f4fk
PubChem CID 9823846
IUPAC Name methanesulfonic acid;2-[4-[(4-nitrophenyl)methoxy]phenyl]ethyl carbamimidothioate
SMILES CS(=O)(=O)O.C1=CC(=CC=C1CCSC(=N)N)OCC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 10 mg
Formula Weight 427.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material S-[4-[(4-Nitrobenzyl)oxy]phenethyl]isothiourea Methanesulfonate
TSCA No
Recommended Storage Refrigerator
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.