Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(-)-5-Iodowillardiine, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

140187-25-3
C7H8IN3O4
325.062
AXXYLTBQIQBTES-BYPYZUCNSA-N
iodo-willardiine,s-5-iodowillardiine,s-5-iodowillardiine,5-iodowillardiine,s---5-iodowillardiine,2-amino-3-5-iodo-2,4-dioxo-3,4-dihydro-2h-pyrimidin-1-yl-propionic acid,iwd,5 iodowillardine,1mqg,tocris-0307
447196
CHEBI:43500
(2S)-2-amino-3-(5-iodo-2,4-dioxopyrimidin-1-yl)propanoic acid
C1=C(C(=O)NC(=O)N1CC(C(=O)O)N)I

Highly potent, subtype-selective kainate agonist

Demonstrates high affinity for the kainate receptor subtype hGluR5 (Ki = 0.24 nM) and 600-4000-fold selectivity over both the AMPA receptor subtypes and the homomeric kainate receptor hGluR6.
Quantity 10mg
Formula Weight 325.06
Percent Purity >98%
Chemical Name or Material (S)-(-)-5-Iodowillardiine

Recommended Storage : Store at Room Temperature