Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ (S)-(-)-Bay K 8644

Catalog No. 154650
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-465-0 50 mg
15-461-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-465-0 Supplier Tocris Bioscience™ Supplier No. 1546/50

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

CaV1.x activator; (S)-(-)-enantiomer of (±)-Bay K 8644

L-type Ca2+-channel activator with positive inotropic, vasoconstrictive and behavioral effects in vivo.

Racemate and (R)-(+)-Enantiomer also available.

Chemical Identifiers

CAS 98625-26-4
Molecular Formula C16H15F3N2O4

Specifications

Quantity 50 mg
Formula Weight 356.3
Purity >99%
Molecular Weight (g/mol) 356.301
Chemical Name or Material (S)-(-)-Bay K 8644
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.