Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(S)-(+)-gamma-Hydroxymethyl-gamma-butyrolactone, 97%

Catalog No. AC337640050
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

Catalog No. AC337640050

Supplier: Thermo Scientific Chemicals 337640050

Only null left
Add to Cart

CAS: 32780-06-6 | C5H8O3 | 116.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32780-06-6
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
MDL Number MFCD00066224
InChI Key NSISJFFVIMQBRN-BYPYZUCNSA-N
Synonym s-5-hydroxymethyl dihydrofuran-2 3h-one, s-+-dihydro-5-hydroxymethyl-2 3h-furanone, s-+-dihydro-5-hydroxymethyl furanone, s-+-gamma-hydroxymethyl-gamma-butyrolactone, 5s-5-hydroxymethyl oxolan-2-one, s-5-hydroxymethyldihydrofuran-2-one, s-5-hydroxymethyl-dihydrofuran-2 3h-one, 2 3h-furanone, dihydro-5-hydroxymethyl-, 5s, s-4-hydroxymethyl butyrolactone, s-+-2',3'-dideoxyribonolactone
PubChem CID 2733813
IUPAC Name (5S)-5-(hydroxymethyl)oxolan-2-one
SMILES C1CC(=O)OC1CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.2300g/mL
Boiling Point 110°C to 115°C (2.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4680 to 1.4720
Quantity 5 g
Specific Gravity 1.23
Specific Rotation Condition +33.5° (20°C 589nm,c=3,EtOH)
Specific Rotation + 33.50
Solubility Information Solubility in water: miscible.
Formula Weight 116.12
Percent Purity 97%
Chemical Name or Material (S)-(+)-γ-Hydroxymethyl-γ-butyrolactone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Avoid breathing dust/fume/

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.