Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SHOWA™ 381 Ultra-Lightweight Microfiber Gloves with Microporous Nitrile Coating
Shop All Showa Best Glove Products
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Description
- Ultra-lightweight gloves designed with flexibility and comfort in mind
- Microfiber construction with embossed microporous nitrile coating
- Thin material ensures optimum tactility when working on delicate tasks
- Breathable microfiber helps reduce palm sweat in warm environments
- Nitrile coating enhances grip capabilities
Specifications
Specifications
| Construction | Seamless Knit |
| Coating Material | Microporous Nitrile |
| Color | Gray, Black |
| Disposable | Reusable |
| Finger Thickness (Metric) | 0.84 mm |
| Hand | Specific |
| Latex Free | Yes |
| Material | Microfiber |
| Product Type | Seamless Knit Gloves |
| Size | Large |
| Show More |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
