Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100 calcium binding protein A14 Antibody (OTI2G9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP273966A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273966A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273966A30 Supplier Novus Biologicals Supplier No. NBP273966AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100 calcium binding protein A14 Monoclonal specifically detects S100 calcium binding protein A14 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen S100 calcium binding protein A14
Applications Western Blot
Classification Monoclonal
Clone OTI2G9
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114
Gene Symbols S100A14
Host Species Mouse
Immunogen Full length recombinant protein of human S100 calcium binding protein A14 (NP_065723) produced in E.coli.
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57402
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.