Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A/B Antibody (6G1cc), Biotin, Novus Biologicals™
SDP

Catalog No. NB001121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001121 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001121 Supplier Novus Biologicals Supplier No. NB200405B0.2ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A/B Monoclonal specifically detects S100A/B in Human samples. It is validated for Western Blot, ELISA, Immunoassay, Sandwich ELISA.

Specifications

Antigen S100A/B
Applications Western Blot, ELISA
Classification Monoclonal
Clone 6G1
Conjugate Biotin
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
Gene Symbols S100A1
Host Species Mouse
Immunogen Human S-100 protein
Quantity 0.2 mL
Research Discipline Breast Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience
Gene ID (Entrez) 6271
Test Specificity This is specific for S-100 beta beta and S-100 alpha beta.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.