Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A/B Antibody (6G1cc), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB359482 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB359482 0.2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB359482 Supplier Novus Biologicals Supplier No. NB200405JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A/B Monoclonal antibody specifically detects S100A/B in Human samples. It is validated for Western Blot, Immunoassay, ELISA, Affinity Purification

Specifications

Antigen S100A/B
Applications Western Blot, Immunoassay, ELISA, Affinity Purification
Classification Monoclonal
Clone 6G1cc
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunoassay, Sandwich ELISA, Immunoaffinity Purification
Formulation 50mM Sodium Borate
Gene Alias protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human brain S100 proteins.
Purification Method Protein A purified
Quantity 0.2 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Biology, Cellular Markers, Neuronal Cell Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 6271
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.