Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A1 Antibody (148), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB074373 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB074373 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB074373 Supplier Novus Biologicals Supplier No. NBP290505G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

S100A1 Monoclonal specifically detects S100A1 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.

Specifications

Antigen S100A1
Applications ELISA
Classification Monoclonal
Clone 148
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A1 (P56565; Gly2-Ser94).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Biology, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6271
Test Specificity No cross-reactivity in ELISA with: Mouse S100B
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.