Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A1 Antibody (OTI6G4), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP273967UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273967UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273967UV Supplier Novus Biologicals Supplier No. NBP273967UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A1 Monoclonal specifically detects S100A1 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen S100A1
Applications Western Blot
Classification Monoclonal
Clone OTI6G4
Conjugate DyLight 350
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
Gene Symbols S100A1
Host Species Mouse
Immunogen Full length recombinant protein of human S100A1 (NP_006262) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Astrocyte Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Hypoxia, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6271.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.