Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A1 Antibody (S100A1/1942), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP259617V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259617V 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259617V Supplier Novus Biologicals Supplier No. NBP259617V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A1 Monoclonal specifically detects S100A1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen S100A1
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S100A1/1942
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias protein S100-A1, S100 alpha, S100 calcium binding protein A1, S100 calcium-binding protein A1S100, S-100 protein alpha chain, S-100 protein subunit alpha, S100 protein, alpha polypeptide, S100A, S100-alpha
Gene Symbols S100A1
Host Species Mouse
Immunogen Recombinant human full-length S100A1 protein (Uniprot: P23297)
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Astrocyte Markers, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Hypoxia, Lipid and Metabolism, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6271
Test Specificity The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and a beta chain whereas S100B is composed of two beta chains. This antibody is specific against an epitope located on the alpha-chain (i.e. in S-100A and S-100B) but not on the beta-chain of S-100 (i.e. in S-100B). This antibody can be used to localize S-100A in various tissue sections. S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle, epithelial and myoepithelial cells of the breast, salivary and sweat glands. Neoplasms derived from these cells also express S-100 protein. Almost all malignant melanomas and cases of histiocytosis X are positive for S-100 protein.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.