Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A10 Antibody (344015), PE/Atto594, Novus Biologicals™
SDP

Catalog No. NB313527 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313527 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313527 Supplier Novus Biologicals Supplier No. FAB2377PEATT594
Add to Cart
Add to Cart

Rat Monoclonal Antibody

S100A10 Monoclonal antibody specifically detects S100A10 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen S100A10
Applications Flow Cytometry
Classification Monoclonal
Clone 344015
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS
Gene Alias annexin II ligand, calpactin I, light polypeptide, ANX2L, ANX2LG, CAL1LGP11,42C, Calpactin I light chain, Calpactin-1 light chain, Cellular ligand of annexin II, CLP11Ca[1], MGC111133, p10, p10 protein, p11, protein S100-A10, S100 calcium binding protein A10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)), S100 calcium-binding protein A10, S100 calcium-binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11))
Host Species Rat
Immunogen E. coli-derived recombinant mouse S100A10, Met1-Lys97, Accession # P08207
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6281
Target Species Mouse
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.