Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A10 Rabbit anti-Human, Clone: 6X8F7, Novus Biologicals™
SDP

Catalog No. NB166030 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166030 100 μg
NB166031 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166030 Supplier Novus Biologicals Supplier No. NBP315467100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

S100A10 Monoclonal antibody specifically detects S100A10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen S100A10
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 6X8F7
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias annexin II ligand, calpactin I, light polypeptide, ANX2L, ANX2LG, CAL1LGP11,42C, Calpactin I light chain, Calpactin-1 light chain, Cellular ligand of annexin II, CLP11Ca[1], MGC111133, p10, p10 protein, p11, protein S100-A10, S100 calcium binding protein A10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)), S100 calcium-binding protein A10, S100 calcium-binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11))
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-97 of human S100A10 (P60903).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 6281
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.