Learn More
Description
Specifications
Specifications
| Antigen | S100A11 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 405 |
| Dilution | Western Blot, ELISA |
| Formulation | 50mM Sodium Borate |
| Gene Alias | calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin) |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant S100A11 (NP_005611.1; Met1-Thr105). |
| Purification Method | Affinity purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
