Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A11 Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB062557 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB062557 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB062557 Supplier Novus Biologicals Supplier No. NBP299718R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S100A11 Polyclonal specifically detects S100A11 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen S100A11
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100CS100 calcium binding protein A11 (calgizzarin)
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant S100A11 (NP_005611.1; Met1-Thr105).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 6282
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.