Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse S100A11 Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | S100A11 |
---|---|
Clone | 493804 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
S100A11 Monoclonal specifically detects S100A11 in Mouse samples. It is validated for Western Blot.Specifications
S100A11 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P50543 | |
6282 | |
E. coli-derived recombinant mouse S100A11 Met1-Ile98 Accession # P50543 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse S100A11 in direct ELISAs and Western blots. In direct ELISAs, ;no cross-reactivity with recombinant mouse S100A1, 4, 6, 8, 9, 10, 13, or 16 is observed, and approximately 75% cross-reactivity with recombinant human (rh) S100A11 is observed. In Western blots, no cross-reactivity with rhS100A11 is observed |
493804 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
calgizzarin, Metastatic lymph node gene 70 protein, MLN 70, MLN70, protein S100-A11, Protein S100-C, S100 calcium binding protein A11, S100 calcium-binding protein A11, S100 calcium-binding protein A11 (calgizzarin), S100C, S100CS100 calcium binding protein A11 (calgizzarin) | |
S100A11 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title