Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A16 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB086109 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB086109 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB086109 Supplier Novus Biologicals Supplier No. NBP299717AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S100A16 Polyclonal specifically detects S100A16 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen S100A16
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50 mM Sodium Borate
Gene Alias Aging-associated gene 13 protein, aging-associated protein 13, DT1P1A7, MGC17528, protein S100-A16, Protein S100-F, S100 calcium binding protein A16, S100FS100 calcium-binding protein A16
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant S100A16 (Q96FQ6; Met 1-Ser 103).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 140576
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.