Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A3 Antibody (001), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB075272 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB075272 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB075272 Supplier Novus Biologicals Supplier No. NBP290583G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

S100A3 Monoclonal specifically detects S100A3 in Mouse samples. It is validated for ELISA.

Specifications

Antigen S100A3
Applications ELISA
Classification Monoclonal
Clone 1
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias Protein S-100E, S100 calcium binding protein A3, S100 calcium-binding protein A3S100Eprotein S100-A3
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A3 (NP_035440.1; Met1-Gln101).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6274
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.