Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A4 Antibody (S100A4/1482), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N25440PE594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N25440PE594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N25440PE594 Supplier Novus Biologicals Supplier No. NBP254400PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A4 Monoclonal specifically detects S100A4 in Human, Mouse samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen S100A4
Applications Flow Cytometry
Classification Monoclonal
Clone S100A4/1482
Conjugate PE/ATTO 594
Dilution Flow Cytometry, Flow (Intracellular)
Formulation PBS with 0.05% Sodium Azide
Gene Alias 18A2, Calvasculin, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog)
Gene Symbols S100A4
Host Species Mouse
Immunogen Recombinant fragment of human S100A4 protein (aa1-200) (exact sequence is proprietary) (Uniprot: P26447)
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6275.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.