Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A4 Antibody (S100A4/2750R), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB392252 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB392252 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB392252 Supplier Novus Biologicals Supplier No. NBP308460APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

S100A4 Monoclonal antibody specifically detects S100A4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen S100A4
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone S100A4/2750R
Conjugate APC
Formulation PBS
Gene Alias 18A2, Calvasculin, CAPLS100 calcium binding protein A4 (calcium protein, calvasculin, metastasin, fibroblast-specific protein-1,42A, FSP1, leukemia multidrug resistance associated protein, malignant transformation suppression 1, Metastasin, MTS1, murine placental homolog), P9KA, PEL98, Placental calcium-binding protein, Protein Mts1, protein S100-A4, S100 calcium binding protein A4, S100 calcium-binding protein A4, S100 calcium-binding protein A4 (calcium protein, calvasculin, metastasin, murine placental homolog)
Host Species Rabbit
Immunogen Recombinant fragment of human S100A4 protein (around aa1-200) (exact sequence is proprietary) (Uniprot: P26447)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6275
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.