Learn More
Description
Specifications
Specifications
| Antigen | S100A5 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50 mM Sodium Borate |
| Gene Alias | Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant S100A5 (P63084; Met 1-Lys 93). |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
