Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A5 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB081701 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB081701 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB081701 Supplier Novus Biologicals Supplier No. NBP298277H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S100A5 Polyclonal specifically detects S100A5 in Mouse samples. It is validated for ELISA.

Specifications

Antigen S100A5
Applications ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Protein S-100D, S100 calcium binding protein A5, S100 calcium-binding protein A5S100Dprotein S100-A5
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant S100A5 (P63084; Met 1-Lys 93).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6276
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.