Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
S100A5 Monoclonal Antibody (C8), Invitrogen™
Mouse Monoclonal Antibody
$243.00 - $522.50
Specifications
Antigen | S100A5 |
---|---|
Clone | C8 |
Concentration | 1 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543596
|
Invitrogen™
MA543596 |
20 μL |
Each for $243.00
|
|
|||||
PIMA543595
|
Invitrogen™
MA543595 |
100 μL |
Each for $522.50
|
|
|||||
Description
Binds calcium, zinc and copper. One subunit can simultaneously bind 2 calcium ions or 2 copper ions plus 1 zinc ion. Calcium and copper ions compete for the same binding sites. Engelkamp D., Proc. Natl. Acad. Sci. U.S.A. 90:6547-6551(1993). Gregory S.G., Nature 441:315-321(2006).Specifications
S100A5 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Protein S100-A5; Protein S-100D; S100 calcium binding protein A5; S100 calcium-binding protein A4; S100 calcium-binding protein A5; S100a5; S100D; S100D9 | |
S100A5 | |
IgG2b κ | |
Protein A/G | |
Antibody |
C8 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P33763 | |
6276 | |
Met1-Lys92 (Accession P33763), with N-terminal His Tag. | |
Primary | |
-20°C | |
S100A5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title