Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A6 Antibody (016), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB039576 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB039576 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB039576 Supplier Novus Biologicals Supplier No. NBP290488G
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

S100A6 Monoclonal specifically detects S100A6 in Mouse samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen S100A6
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 16
Conjugate DyLight 488
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 2A9, CABP, CACY5B10, calcyclin, Growth factor-inducible protein 2A9, MLN 4, PRAS100 calcium binding protein A6 (calcyclin), Prolactin receptor-associated protein, protein S100-A6, S100 calcium binding protein A6, S100 calcium-binding protein A6, S100 calcium-binding protein A6 (calcyclin)
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Mouse S100A6 (Met 1-Lys 89; NP_035443.1).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 6277
Test Specificity No cross-reactivity in ELISA with: Human S100A1 Human S100A2/ CAN19/ S100L
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.