Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A7/Psoriasin Antibody (47C1068) - Azide Free, Novus Biologicals™
SDP

Catalog No. NBP224911 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224911 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224911 Supplier Novus Biologicals Supplier No. NBP224911
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 14 publications

S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.

Specifications

Antigen S100A7/Psoriasin
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Classification Monoclonal
Clone 47C1068
Concentration 0.5mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, Simple Western 1:2000, Flow Cytometry, Immunohistochemistry reported in scientific literature (PMID 27328057), Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready
Gene Accession No. P31151
Gene Alias HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c
Gene Symbols S100A7
Host Species Mouse
Immunogen This antibody was developed against recombinant psoriasin/HID-5 protein.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6278
Test Specificity In confluent MCF10A cells, an 11 kDa band is observed.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.