Learn More
Description
Specifications
Specifications
| Antigen | S100A7/Psoriasin |
| Applications | Electron Microscopy, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Monoclonal |
| Clone | 47C1068 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1-3 ug/ml, Simple Western 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 26225121), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy reported in scientific literature (PMID 26225121), Flow (Intracellular) |
| Gene Accession No. | P31151 |
| Gene Alias | HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c |
| Gene Symbols | S100A7 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
