Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A7/Psoriasin Antibody (47C1068) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody has been used in 47 publications

$206.50 - $501.50

Specifications

Antigen S100A7/Psoriasin
Clone 47C1068
Concentration 1.0 mg/mL
Dilution Western Blot 1-3 ug/ml, Simple Western 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 26225121), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy reported in scientific literature (PMID 26225121), Flow (Intracellular)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB10056559T
SDP
View Documents
Novus Biologicals
NB10056559SS
0.025 mg
Each for $206.50
Only null left
Add to Cart
 
NB10056559
SDP
View Documents
Novus Biologicals
NB10056559
0.1 mg
Each for $501.50
Only null left
Add to Cart
 
Description

Description

S100A7/Psoriasin Monoclonal specifically detects S100A7/Psoriasin in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy, Flow (Intracellular).
Specifications

Specifications

S100A7/Psoriasin
1.0 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoassay, Flow Cytometry
Unconjugated
Mouse
Human, Mouse
HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c
S100A7
IgG1 κ
Protein G purified
In confluent MCF10A cells, an 11 kDa band is observed.
47C1068
Western Blot 1-3 ug/ml, Simple Western 1:500, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation reported in scientific literature (PMID 26225121), Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Electron Microscopy reported in scientific literature (PMID 26225121), Flow (Intracellular)
Monoclonal
Purified
RUO
P31151
6278
This antibody was developed against recombinant psoriasin/HID-5 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.