Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A7/Psoriasin Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB323958 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB323958 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB323958 Supplier Novus Biologicals Supplier No. NBP299720PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

S100A7/Psoriasin Polyclonal antibody specifically detects S100A7/Psoriasin in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen S100A7/Psoriasin
Applications Western Blot, ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Western Blot, ELISA, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias HID 5, HID5, HID-5, protein S100-A7, PSOR1, Psoriasin, psoriasin 1, S100 calcium binding protein A7, S100 calcium binding protein A7 (psoriasin 1), S100 calcium-binding protein A7, S100 calcium-binding protein A7 (psoriasin 1), S100A7c
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human S100A7/Psoriasin (Accession#: NP_002954.2; Met 1-Gln 101)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6278
Target Species Human, Mouse (Negative), Rat (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.