Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/A9 Antibody (48M7C7), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. N225269J549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N225269J549 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N225269J549 Supplier Novus Biologicals Supplier No. NBP225269JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A8/A9 Monoclonal specifically detects S100A8/A9 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen S100A8/A9
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 48M7C7
Conjugate Janelia Fluor 549
Dilution Western Blot, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8, S100 calcium binding protein A8, S100A8
Gene Symbols S100A8
Host Species Mouse
Immunogen Within the range of amino acids 2-105 of human S100A8 were used as the immunogen for the antibody.
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6279.0
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.