Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/A9 Antibody (5.5), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP250283AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250283AI 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250283AI Supplier Novus Biologicals Supplier No. NBP250283AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A8/A9 Monoclonal specifically detects S100A8/A9 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.

Specifications

Antigen S100A8/A9
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5.5
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 60B8AG, CAGA, CFAG, CGLA, CP-10, L1Ag, MA387, MIF, MRP8, NIF, P8, S100 calcium binding protein A8, S100A8
Gene Symbols S100A8
Host Species Mouse
Immunogen Acute monocytic leukaemia cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6279
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.