Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Antibody (236), Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB109405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB109405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB109405 Supplier Novus Biologicals Supplier No. NBP289809AF488
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

S100A8 Monoclonal specifically detects S100A8 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen S100A8
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 236
Conjugate Alexa Fluor 488
Formulation 50 mM Sodium Borate
Gene Alias 60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human S100A8 (NP_002955.2; Met1-Glu93).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6279
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.