Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8 Antibody (2C5/4), DyLight 594, Novus Biologicals™
SDP

Catalog No. NP27660DL94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP27660DL94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP27660DL94 Supplier Novus Biologicals Supplier No. NBP276650DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A8 Monoclonal specifically detects S100A8 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen S100A8
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2C5/4
Conjugate DyLight 594
Dilution Western Blot, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias 60B8AG, CAGACP-10, calgranulin A, calgranulin-A, Calprotectin L1L subunit, CFAGL1Ag, CGLA, Cystic fibrosis antigen, Leukocyte L1 complex light chain, MA387, MIF, Migration inhibitory factor-related protein 8, MRP-8, MRP8S100 calcium binding protein A8 (calgranulin A), NIF, P8, protein S100-A8, S100 calcium binding protein A8, S100 calcium-binding protein A8, S100 calcium-binding protein A8 (calgranulin A), Urinary stone protein band A
Gene Symbols S100A8
Host Species Mouse
Immunogen Recombinant human S100A8
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6279
Test Specificity This antibody is specific for S100A8 both as an isolated protein and in combination with S100A9.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.