Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/S100A9 Heterodimer Antibody (900027), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NB303526 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB303526 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB303526 Supplier Novus Biologicals Supplier No. FAB45702N

Mouse Monoclonal Antibody

S100A8/S100A9 Heterodimer Monoclonal antibody specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for ELISA, Immunohistochemistry, Flow Cytometry, CyTOF

Specifications

Antigen S100A8/S100A9 Heterodimer
Applications ELISA, Immunohistochemistry, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 900027
Conjugate Alexa Fluor 700
Dilution ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) and Thr2-Pro114 (S100A9), Accession # P05109 (S100A8) and P06702 (S100A9)
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.