Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/S100A9 Heterodimer Antibody (900028), Biotin, Novus Biologicals™
SDP

Catalog No. NB369814 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369814 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369814 Supplier Novus Biologicals Supplier No. FAB45703B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A8/S100A9 Heterodimer Monoclonal specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry (Intracellular Staining).

Specifications

Antigen S100A8/S100A9 Heterodimer
Applications CyTOF, ELISA, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 900028
Conjugate Biotin
Dilution ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Host Species Mouse
Immunogen E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9), Accession # P05109 (S100A8) and P06702 (S100A9)
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.