Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A8/S100A9 Heterodimer Antibody (900028), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347772 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347772 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347772 Supplier Novus Biologicals Supplier No. FAB45703MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A8/S100A9 Heterodimer Monoclonal specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for CyTOF, ELISA, Flow Cytometry (Intracellular Staining).

Specifications

Antigen S100A8/S100A9 Heterodimer
Applications CyTOF, ELISA, Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone 900028
Conjugate mFluor Violet 610 SE
Dilution ELISA, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen E. coli-derived recombinant human S100A8/S100A9 heterodimer, Met1-Glu93 (S100A8) and Thr2-Pro114 (S100A9), Accession # P05109 (S100A8) and P06702 (S100A9)
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.