Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human S100A8/S100A9 Heterodimer Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$214.00 - $526.00
Specifications
Antigen | S100A8/S100A9 Heterodimer |
---|---|
Clone | 1099F |
Dilution | Immunohistochemistry 10-25 ug/mL, Immunocytochemistry 3-25 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL |
Applications | Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH) |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB45701SP
![]() |
R&D Systems
MAB45701SP |
25 μg |
Each for $214.00
|
|
|||||
MAB45701100
![]() |
R&D Systems
MAB45701100 |
100 μg |
Each for $526.00
|
|
|||||
Description
S100A8/S100A9 Heterodimer Monoclonal specifically detects S100A8/S100A9 Heterodimer in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry, Dual RNAscope ISH-IHC.Specifications
S100A8/S100A9 Heterodimer | |
Immunohistochemistry 10-25 ug/mL, Immunocytochemistry 3-25 ug/mL, Dual RNAscope ISH-IHC 5-25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Supplied as a solution in PBS containing BSA, Glycerol and Sodium Azide. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with Sodium Azide | |
E. coli-derived recombinant human S100A8/S100A9 heterodimer Met1-Glu93 (S100A8) & Thr2-Pro114 (S100A9) Accession # P05109 (S100A8) and P06702 (S100A9) | |
Primary | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
1099F | |
Immunohistochemistry, Immunocytochemistry, In Situ Hybridization (ISH) | |
Unconjugated | |
Rabbit | |
Human | |
P05109 (S100A8) and P06702 (S100A9) | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Detects human S100A8/S100A9 in direct ELISAs. In ELISAs, it detects recombinant human S100A8/S100A9 heterodimer but it does not detect recombinant human S100A8 or S100A9 monomers. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title