Learn More
Description
Specifications
Specifications
| Antigen | S100A9 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 25792748), Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin reported in scientific literature (PMID 21653680) |
| Formulation | Whole antisera with 0.1% Sodium Azide |
| Gene Accession No. | P06702 |
| Gene Alias | CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B) |
| Gene Symbols | S100A9 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
