Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A9 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 13 publications

$256.00 - $512.00

Specifications

Antigen S100A9
Dilution Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 25792748), Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin reported in scientific literature (PMID 21653680)
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11089726T
SDP
View Documents
Novus Biologicals
NB11089726SS
0.025 mL
Each for $256.00
Only null left
Add to Cart
 
NB11089726
SDP
View Documents
Novus Biologicals
NB11089726
0.1 mL
Each for $512.00
Only null left
Add to Cart
 
Description

Description

S100A9 Polyclonal specifically detects S100A9 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

S100A9
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases
Whole antisera with 0.1% Sodium Azide
CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
S100A9
IgG
Unpurified
16 kDa
Western Blot 1:1000, Simple Western 1:100, Immunohistochemistry reported in scientific literature (PMID 25792748), Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin reported in scientific literature (PMID 21653680)
Polyclonal
Antisera
RUO
Human, Mouse, Rat
P06702
6280
Full length human S100A9 protein [Swiss-Prot# P06702] expressed in E. coli.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.