Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A9 Antibody (OTI10D5) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP273970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273970 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273970 Supplier Novus Biologicals Supplier No. NBP273970
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A9 Monoclonal specifically detects S100A9 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen S100A9
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI10D5
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunohistochemistry 1:150
Formulation Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
Gene Alias CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
Gene Symbols S100A9
Host Species Mouse
Immunogen Full length human recombinant protein of human S100A9 (NP_002956) produced in SF9 cell.
Molecular Weight of Antigen 13.1 kDa
Purification Method Affinity Purified
Quantity 100 μg
Research Discipline Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6280
Reconstitution Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.