Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100A9 Antibody (S100A9/1011), Novus Biologicals™
SDP

Catalog No. NBP24491001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP24491001 0.1 mg
NBP24491000 0.02 mg
NBP24491002 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP24491001 Supplier Novus Biologicals Supplier No. NBP2449100.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100A9 Monoclonal specifically detects S100A9 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen S100A9
Applications Immunohistochemistry
Classification Monoclonal
Clone S100A9/1011
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1 ug/ml, Immunohistochemistry-Frozen (Negative), Immunofluorescence 0.5 - 1.0 ug/ml
Formulation 10mM PBS with 0.05% Sodium Azide
Gene Alias CAGBMigration inhibitory factor-related protein 14, calgranulin B, calgranulin-B, Calprotectin L1H subunit, CFAGMRP-14,60B8AG, CGLB, L1AG, LIAG, MAC387, MIF, MRP14Leukocyte L1 complex heavy chain, NIF, P14, protein S100-A9, S100 calcium binding protein A9, S100 calcium binding protein A9 (calgranulin B), S100 calcium-binding protein A9, S100 calcium-binding protein A9 (calgranulin B)
Gene Symbols S100A9
Host Species Mouse
Immunogen Recombinant human S100A9 protein
Molecular Weight of Antigen 14 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity, Signal Transduction, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 6280
Test Specificity Recognizes a 14kDa protein, identified as S100A9 (also known as Calgranulin B or MRP-14); expressed by granulocytes, monocytes and by tissue macrophages.The protein encoded by this gene is a member of the S100 family of proteins containing 2 EF-hand calcium-binding motifs. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells, and involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. Altered expression of this protein is associated with the disease cystic fibrosis. This MAb reacts with neutrophils, monocytes and macrophages, and has been shown as an important marker for identifying macrophages in tissue sections. Among cells that are now recognized as macrophages are histiocytes, Kupffer cells, osteoclasts, microglial cells, synovial type A cells, interdigitating cells, and Langerhans cells (in normal tissues) and epithelioid cells and Langerhans-type and foreign-body-type multinucleated giant cells (in inflamed tissues).
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.