Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Antibody (S1-61), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP250282AJ Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250282AJ 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250282AJ Supplier Novus Biologicals Supplier No. NBP250282AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100B Monoclonal specifically detects S100B in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen S100B
Applications Immunohistochemistry
Classification Monoclonal
Clone S1-61
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Gene Symbols S100B
Host Species Mouse
Immunogen S100B protein conjugated to methylated BSA
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6285
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.