Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Antibody (S1-61), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB309641 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309641 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309641 Supplier Novus Biologicals Supplier No. NBP250282MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

S100B Monoclonal antibody specifically detects S100B in Human samples. It is validated for Immunohistochemistry

Specifications

Antigen S100B
Applications Immunohistochemistry
Classification Monoclonal
Clone S1-61
Conjugate mFluor Violet 610 SE
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Host Species Mouse
Immunogen S100B protein conjugated to methylated BSA
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6285
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.