| Antigen |
S100B |
| Applications |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification |
Monoclonal |
| Clone |
S100B/1012 |
| Conjugate |
DyLight 550 |
| Dilution |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Formulation |
50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias |
beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta |
| Gene Symbols |
S100B |
| Host Species |
Mouse |
| Immunogen |
Recombinant full-length human S100B protein (Uniprot: P04271) |
| Purification Method |
Protein A or G purified |
| Quantity |
100 μL |
| Regulatory Status |
RUO |
| Research Discipline |
Alzheimers Research, Apoptosis, Biologically Active Proteins, Breast Cancer, Cancer, Cell Biology, Cellular Markers, Epigenetics, Hypoxia, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Peroxisome Markers, Signal Transduction, Stem Cells |
| Primary or Secondary |
Primary |
| Gene ID (Entrez) |
6285 |
| Test Specificity |
The specificity of this monoclonal antibody to its intended target was validated by HuProtTM Array, containing more than 19,000, full-length human proteins. S100 belongs to the family of calcium binding proteins. S100A and S100B proteins are two members of the S100 family. S100A is composed of an alpha and a beta chain whereas S100B is composed of two beta chains. This antibody is specific against an epitope located on the beta-chain (i.e. in S-100A and S-100B) but not on the alpha-chain of S-100 (i.e. in S-100A and S100A0). This antibody can be used to localize S-100A and S-100B in various tissue sections. S-100 protein has been found in normal melanocytes, Langerhans cells, histiocytes, chondrocytes, lipocytes, skeletal and cardiac muscle, Schwann cells, epithelial and myoepithelial cells of the breast, salivary and sweat glands, as well as in glial cells. Neoplasms derived from these cells also express S-100 protein, albeit non-uniformly. A large number of well-differentiated tumors of the salivary gland, adipose and cartilaginous tissue, and Schwann cell-derived tumors express S-100 protein. Almost all malignant melanomas and cases of histiocytosis X are positive for S-100 protein. |
| Target Species |
Human, Mouse, Rat, Bovine |
| Content And Storage |
Store at 4C in the dark. |
| Form |
Purified |
| Isotype |
IgG2a κ |
|
Show More
Show Less
|
|