Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

S100B Mouse anti-Bovine, Human, Mouse, Rat, Clone-4C4.9, Novus Biologicals™
SDP

Catalog No. NB254579200 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB254579200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB254579200 Supplier Novus Biologicals Supplier No. NBP254579200UG

Mouse Monoclonal Antibody

IHC-P: Staining of formalin-fixed tissues requires boiling tissue sections in 10mM citrate buffer pH 6.0 for 10-20 min followed by cooling at RT for 20 minutes

Specifications

Antigen S100B
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone 4C4.9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5 - 1 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 0.25 - 0.5 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias beta (neural), NEF, S100, S100 beta, S100 calcium binding protein B, S100 calcium-binding protein B, S100 calcium-binding protein, beta (neural), S-100 calcium-binding protein, beta chain, 10protein S100-B, S-100 protein beta chain, S-100 protein subunit beta, S100beta
Gene Symbols S100B
Host Species Mouse
Immunogen Purified bovine brain S100 protein
Molecular Weight of Antigen 11 kDa
Purification Method Protein A or G purified
Quantity 200 μg
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cytoskeleton Markers, Neuroscience, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6285
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at -20 to -80C. Avoid freeze-thaw cycles.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.